accessibility menu, dialog, popup

UserName

CH3OHH3C

CAS RN 20739-59-7

IUPAC Name: hex-4-yn-3-ol
Synonyms: hex-4-yn-3-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: KARLLBDFLHNKBO-UHFFFAOYNA-N
SMILES: CCC(O)C#CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)