accessibility menu, dialog, popup

UserName

OPOOONaNa

CAS RN 207569-06-0

IUPAC Name: disodium hydrate naphthalen-1-yl phosphate
Synonyms: disodium hydrate naphthalen-1-yl phosphate
Molecular Weight (g/mol): 286.13
Molecular Formula: C10H9Na2O5P
InChi Key: RJJPCVOIUDUWLF-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC2=CC=CC=C12
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results