accessibility menu, dialog, popup

UserName

CAS RN 20769-85-1

CH3H3CBrBrO

CAS RN 20769-85-1

IUPAC Name: 2-bromo-2-methylpropanoyl bromide
Synonyms: 2-bromo-2-methylpropanoyl bromide
Molecular Weight (g/mol): 229.9
Molecular Formula: C4H6Br2O
InChi Key: YOCIJWAHRAJQFT-UHFFFAOYSA-N
SMILES: CC(C)(Br)C(Br)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)