accessibility menu, dialog, popup

UserName

O(S)

CAS RN 20780-54-5

IUPAC Name: (2S)-2-phenyloxirane
Synonyms: (S)-styrene oxide S-(epoxyethyl)benzene S-phenyloxirane
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: AWMVMTVKBNGEAK-MRVPVSSYSA-N
SMILES: C1O[C@H]1C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results