accessibility menu, dialog, popup

UserName

IHNOO

CAS RN 20780-76-1

IUPAC Name: 5-iodo-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-iodo-1H-indole-2,3-dione
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H4INO2
InChi Key: OEUGDMOJQQLVAZ-UHFFFAOYSA-N
SMILES: IC1=CC=C2NC(=O)C(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results