accessibility menu, dialog, popup

UserName

CH3SOONa

CAS RN 207801-20-5

IUPAC Name: 4-methylbenzene-1-sulfinate
Synonyms: 4-methylbenzenesulfinate
Molecular Weight (g/mol): 155.19
Molecular Formula: C7H7O2S
InChi Key: FXJVNINSOKCNJP-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results