accessibility menu, dialog, popup

UserName

CAS RN 20782-94-9

CH3ClCH3NH2

CAS RN 20782-94-9

IUPAC Name: 4-chloro-2,5-dimethylaniline
Synonyms: 4-chloro-2,5-dimethylaniline
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: POEANWWEPFQEEF-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(C)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results