accessibility menu, dialog, popup

UserName

CAS RN 208663-08-5

CH3ClHNOOOH(E/Z)

CAS RN 208663-08-5

IUPAC Name: 3-[(5-chloro-2-methylphenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(5-chloro-2-methylphenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 239.66
Molecular Formula: C11H10ClNO3
InChi Key: KFLFGXYJSLVOCF-UHFFFAOYSA-N
SMILES: CC1=C(NC(=O)C=CC(O)=O)C=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results