accessibility menu, dialog, popup

UserName

BrHNO

CAS RN 20870-78-4

IUPAC Name: 5-bromo-2,3-dihydro-1H-indol-2-one
Synonyms: 5-bromo-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: VIMNAEVMZXIKFL-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)CC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results