accessibility menu, dialog, popup

UserName

H3CNOBr

CAS RN 20870-90-0

IUPAC Name: 5-bromo-1-methyl-2,3-dihydro-1H-indol-2-one
Synonyms: 5-bromo-1-methyl-3H-indol-2-one
Molecular Weight (g/mol): 226.07
Molecular Formula: C9H8BrNO
InChi Key: WARSUKBSFLACOI-UHFFFAOYSA-N
SMILES: CN1C(=O)CC2=CC(Br)=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results