accessibility menu, dialog, popup

UserName

CAS RN 20986-40-7

H3COONBr

CAS RN 20986-40-7

IUPAC Name: ethyl 5-bromopyridine-3-carboxylate
Synonyms: ethyl 5-bromopyridine-3-carboxylate
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: PCPIANOJERKFJI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=CC(Br)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results