CAS RN 2104-19-0
CAS RN 2104-19-0
IUPAC Name:
9-methoxy-9-oxononanoic acid
Synonyms:
9-methoxy-9-oxononanoic acid
Molecular Weight (g/mol):
202.25
Molecular Formula:
C10H18O4
InChi Key:
VVWPSAPZUZXYCM-UHFFFAOYSA-N
SMILES:
COC(=O)CCCCCCCC(O)=O
1
–
2
of
2
results
1
8-Methoxycarbonyloctanoic Acid, TRC
CAS: 2104-19-0 Molecular Formula: C10 H18 O4 Molecular Weight (g/mol): 202.25 Synonym: Azelaic Acid Monomethyl Ester IUPAC Name: 9-methoxy-9-oxononanoic acid SMILES: COC(=O)CCCCCCCC(=O)O
2
Methyl hydrogen azelate, 96%
CAS: 2104-19-0 Molecular Formula: C10H18O4 Molecular Weight (g/mol): 202.25 MDL Number: MFCD00004431 InChI Key: VVWPSAPZUZXYCM-UHFFFAOYSA-N Synonym: methyl hydrogen azelate,monomethyl azelate,azelaic acid monomethyl ester,nonanedioic acid, monomethyl ester,monomethyl nonanedioate,8-carbomethoxyoctanoic acid,azelaic acid, monomethyl ester,mono-methyl azelate,unii-xdi67hf5bh,xdi67hf5bh PubChem CID: 75009 IUPAC Name: 9-methoxy-9-oxononanoic acid SMILES: COC(=O)CCCCCCCC(=O)O