accessibility menu, dialog, popup

UserName

CAS RN 211029-67-3

IUPAC Name: tert-butyl N-(3-iodopyridin-4-yl)carbamate
Synonyms: tert-butyl N-(3-iodopyridin-4-yl)carbamate
Molecular Weight (g/mol): 320.13
Molecular Formula: C10H13IN2O2
InChi Key: FFJIDTLUJQCVAI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=C(I)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results