accessibility menu, dialog, popup

UserName

CAS RN 21168-71-8

H3CNNH2

CAS RN 21168-71-8

IUPAC Name: 1-(1-ethylpiperidin-4-yl)methanamine
Synonyms: 1-(1-ethylpiperidin-4-yl)methanamine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: ZLODGCYXZYPQKQ-UHFFFAOYSA-N
SMILES: CCN1CCC(CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results