CAS RN 21233-61-4
CAS RN 21233-61-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl quinoline-4-carboxylate (~90%), TRC
CAS: 21233-61-4 Molecular Formula: C11H9NO2 Molecular Weight (g/mol): 187.19 Synonym: Methyl quinoline-4-carboxylate IUPAC Name: methyl quinoline-4-carboxylate SMILES: COC(=O)c1ccnc2ccccc12