accessibility menu, dialog, popup

UserName

BOCH3FFHOOH

CAS RN 212386-71-5

IUPAC Name: (4-ethoxy-2,3-difluorophenyl)boronic acid
Synonyms: 4-ethoxy-2,3-difluorophenylboronic acid
Molecular Weight (g/mol): 201.96
Molecular Formula: C8H9BF2O3
InChi Key: OBKSFBWOZSQGGC-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(F)=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results