accessibility menu, dialog, popup

UserName

H3CONCS

CAS RN 2131-57-9

IUPAC Name: 1-(4-isothiocyanatophenyl)ethan-1-one
Synonyms: 1-(4-isothiocyanatophenyl)ethanone
Molecular Weight (g/mol): 177.22
Molecular Formula: C9H7NOS
InChi Key: KAKXOESNHIUYJT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results