accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHSBr

CAS RN 215183-27-0

IUPAC Name: tert-butyl N-[(5-bromothiophen-2-yl)methyl]carbamate
Synonyms: tert-butyl N-[(5-bromothiophen-2-yl)methyl]carbamate
Molecular Weight (g/mol): 292.19
Molecular Formula: C10H14BrNO2S
InChi Key: CVNITIPQHIGCKI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results