CAS RN 2154342-56-8
CAS RN 2154342-56-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Pomalidomide 4'-PEG1-amine
Cereblon ligand with PEG linker and terminal amine for onward chemistry