accessibility menu, dialog, popup

UserName

FBrIFFBr

CAS RN 216394-01-3

IUPAC Name: 1,2-dibromo-1,1,2-trifluoro-2-iodoethane
Synonyms: 1,2-dibromo-1,1,2-trifluoro-2-iodoethane
Molecular Weight (g/mol): 367.73
Molecular Formula: C2Br2F3I
InChi Key: YUBJHLYGOOZDRN-UHFFFAOYNA-N
SMILES: FC(F)(Br)C(F)(Br)I
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results