accessibility menu, dialog, popup

UserName

BCH3FFFK

CAS RN 216434-82-1

IUPAC Name: potassium trifluoro(4-methylphenyl)boranuide
Synonyms: potassium trifluoro(4-methylphenyl)boranuide
Molecular Weight (g/mol): 198.04
Molecular Formula: C7H7BF3K
InChi Key: KRWDYXJWQBTBAH-UHFFFAOYSA-N
SMILES: [K+].CC1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results