accessibility menu, dialog, popup

UserName

CAS RN 21645-51-2

IUPAC Name: alumanetriol
Synonyms: aluminium hydroxide
Molecular Weight (g/mol): 81.03
Molecular Formula: AlH6O3
InChi Key: MXRIRQGCELJRSN-UHFFFAOYSA-N
SMILES: O.O.O.[Al]
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
Grade 
  • (4)
  • (6)
Physical Form 
  • (3)
  • (4)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (10)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (6)
  • (3)
  • (4)
  • (4)