accessibility menu, dialog, popup

UserName

CAS RN 21651-19-4

IUPAC Name: stannanone
Synonyms: tin(II) oxide
Molecular Weight (g/mol): 134.71
Molecular Formula: OSn
InChi Key: QHGNHLZPVBIIPX-UHFFFAOYSA-N
SMILES: O=[Sn]
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results