accessibility menu, dialog, popup

UserName

CAS RN 21652-58-4

FFFFFFFFFFFFFFFFFH2C

CAS RN 21652-58-4

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene
Synonyms: tridecafluoro 1-decene
Molecular Weight (g/mol): 446.11
Molecular Formula: C10H3F17
InChi Key: NKAMGQZDVMQEJL-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)