accessibility menu, dialog, popup

UserName

CAS RN 21714-25-0

IUPAC Name: 4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
Synonyms: 4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
Molecular Weight (g/mol): 223.29
Molecular Formula: C11H13NO2S
InChi Key: GHYFLDWHIVKQLS-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCN=C=S)C=C1OC
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results