CAS RN 21740-23-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,5-Di-O-(p-chlorobenzoyl)-2-deoxy-α-D-ribofuranosyl Chloride, TRC
CAS: 21740-23-8 Molecular Formula: C19 H15 Cl3 O5 Molecular Weight (g/mol): 429.6784 Synonym: 2-Deoxy-α-D-erythro-pentofuranosyl Chloride 3,5-Bis(4-chlorobenzoate),1-Chloro-2-deoxy-3,5-O-bis(p-chlorobenzoyl)-α-D-erythro-pentofuranose,3,5-Bis(4-chlorobenzoyl)-2-deoxy-.alpha.-D-ribofuranosyl Chloride,NSC 140594,3,5-Di-O-(p-chlorobenzoyl)-2-deoxy-α-D-ribofuranosyl Chloride IUPAC Name: [(2R,3S)-5-chloro-3-(4-chlorobenzoyl)oxyoxolan-2-yl]methyl 4-chlorobenzoate SMILES: ClC1C[C@H](OC(=O)c2ccc(Cl)cc2)[C@@H](COC(=O)c3ccc(Cl)cc3)O1