accessibility menu, dialog, popup

UserName

CAS RN 2177-47-1

H3C

CAS RN 2177-47-1

IUPAC Name: 2-methyl-1H-indene
Synonyms: 2-methyl-1H-indene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: YSAXEHWHSLANOM-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results