accessibility menu, dialog, popup

UserName

H3CNNOHO

CAS RN 21801-79-6

IUPAC Name: 2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Synonyms: 2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: FBIUGCLQMKPURJ-UHFFFAOYSA-N
SMILES: CC1=C(N2C=CC=CC2=N1)C(O)=O

 No Refinements