accessibility menu, dialog, popup

UserName

CAS RN 2192-06-5

IUPAC Name: 2-amino-4,5-dihydro-1,3-thiazol-4-one hydrochloridyl
Synonyms: 4(5H)-thiazolone, 2-amino- hydrochloridyl
Molecular Weight (g/mol): 151.59
Molecular Formula: C3H4ClN2OS
InChi Key: CJPDNRMSRPXGQO-UHFFFAOYSA-N
SMILES: [Cl].NC1=NC(=O)CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results