accessibility menu, dialog, popup

UserName

CAS RN 22007-68-7

IUPAC Name: 2-methylpropanimidamide hydrochloridyl
Synonyms: 2-methylpropanimidamide hydrochloridyl
Molecular Weight (g/mol): 121.59
Molecular Formula: C4H10ClN2
InChi Key: AMRZZOYMVJYCNM-UHFFFAOYSA-N
SMILES: [Cl].CC(C)C(N)=N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results