accessibility menu, dialog, popup

UserName

CAS RN 22090-24-0

OON

CAS RN 22090-24-0

IUPAC Name: 4-(piperidin-1-yl)benzoate
Synonyms: 4-(piperidin-1-yl)benzoate
Molecular Weight (g/mol): 204.25
Molecular Formula: C12H14NO2
InChi Key: DGJNUESQBVPXAY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results