accessibility menu, dialog, popup

UserName

CAS RN 2212-75-1

H2N(S)NHOOOHO

CAS RN 2212-75-1

IUPAC Name: (2S)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms: (2S)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: OJTJKAUNOLVMDX-LBPRGKRZSA-N
SMILES: NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)