CAS RN 22144-77-0
CAS RN 22144-77-0
IUPAC Name:
(3S,3aR,4S,6S,6aR,10S,12R,15R,15aR)-3-benzyl-6,12-dihydroxy-4,10,12-trimethyl-5-methylidene-1,11-dioxo-1H,2H,3H,3aH,4H,5H,6H,6aH,9H,10H,11H,12H,15H-cycloundeca[e]isoindol-15-yl acetate
Synonyms:
cytochalasin D
Molecular Weight (g/mol):
507.63
Molecular Formula:
C30H37NO6
InChi Key:
SDZRWUKZFQQKKV-JHADDHBZSA-N
SMILES:
C[C@H]1[C@H]2[C@H](CC3=CC=CC=C3)NC(=O)[C@@]22[C@@H](\C=C\C[C@H](C)C(=O)[C@](C)(O)\C=C\[C@H]2OC(C)=O)[C@H](O)C1=C
1
–
2
of
2
results
1
Cytochalasin D, 98.2%, MP Biomedicals™
CAS: 22144-77-0 Molecular Formula: C30H37NO6 Molecular Weight (g/mol): 507.627 InChI Key: SDZRWUKZFQQKKV-DKLHZCHASA-N Synonym: cytochalasin d PubChem CID: 131668624 SMILES: CC1CC=CC2C(C(=C)C(C3C2(C(C=CC(C1=O)(C)O)OC(=O)C)C(=O)NC3CC4=CC=CC=C4)C)O
2
Tocris Bioscience™ Cytochalasin D
CAS: 22144-77-0 Molecular Formula: C30H37NO6