CAS RN 222036-27-3
CAS RN 222036-27-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6,8-Dihydro-7H-pyrrolo[2,3-g]benzothiazol-7-one, TRC
CAS: 222036-27-3 Molecular Formula: C9H6N2OS Molecular Weight (g/mol): 190.22 Synonym: 6H-Thiazolo[5,4-e]indol-7(8H)-one SMILES: O=C1Cc2c(N1)ccc3ncsc23