CAS RN 2227-57-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(4-Methoxyphenyl)-2-propynoic Acid, TRC
CAS: 2227-57-8 Molecular Formula: C10H8O3 Molecular Weight (g/mol): 176.17 Synonym: 3-(4-Methoxyphenyl)propiolic acid IUPAC Name: 3-(4-methoxyphenyl)prop-2-ynoic acid SMILES: COc1ccc(cc1)C#CC(=O)O