accessibility menu, dialog, popup

UserName

CAS RN 22282-69-5

IUPAC Name: 2-fluoro-1,4-dihydropyridin-4-one
Synonyms: 2-fluoro-1H-pyridin-4-one
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: BRLSDLHMGRDAPF-UHFFFAOYSA-N
SMILES: FC1=CC(=O)C=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results