accessibility menu, dialog, popup

UserName

CAS RN 22288-78-4

CH3OOSH2N

CAS RN 22288-78-4

IUPAC Name: methyl 3-aminothiophene-2-carboxylate
Synonyms: methyl 3-aminothiophene-2-carboxylate
Molecular Weight (g/mol): 157.19
Molecular Formula: C6H7NO2S
InChi Key: TWEQNZZOOFKOER-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=CS1
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)