accessibility menu, dialog, popup

UserName

CAS RN 2243-73-4

IUPAC Name: 3-nitro-N-phenylbenzamide
Synonyms: 3-nitro-N-phenylbenzamide
Molecular Weight (g/mol): 242.23
Molecular Formula: C13H10N2O3
InChi Key: VWIRWLAPFZXYSL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results