accessibility menu, dialog, popup

UserName

CAS RN 2243-82-5

NH2O

CAS RN 2243-82-5

IUPAC Name: naphthalene-2-carboxamide
Synonyms: 2naphthylamide
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: JVXXKQIRGQDWOJ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results