CAS RN 22439-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dibenzothiophene-2-ol, TRC
CAS: 22439-65-2 Molecular Formula: C12H8OS Molecular Weight (g/mol): 200.26 Synonym: 2-Hydroxydibenzothiophene,3-Hydroxydiphenylenesulfide,Benzo[b]benzo[b]thiophen-2-ol; SMILES: Oc1ccc2sc3ccccc3c2c1