accessibility menu, dialog, popup

UserName

LiH3CCH3H3COHCOCH3CH3H3C

CAS RN 22441-13-0

IUPAC Name: lithium(1+) 2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide
Synonyms: lithium(1+) 2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide
Molecular Weight (g/mol): 190.21
Molecular Formula: C11H19LiO2
InChi Key: JUZHUMQSMMHACA-UHFFFAOYSA-N
SMILES: [Li+].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results