CAS RN 2251047-36-4
CAS RN 2251047-36-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Methylpyridin-2-one Pirfenidone, TRC
CAS: 2251047-36-4 Molecular Formula: C18H16N2O2 Molecular Weight (g/mol): 292.33 Synonym: 1,1'-(1,4-Phenylene)bis[5-methyl-2(1H)-pyridinone IUPAC Name: 5-methyl-1-[4-(5-methyl-2-oxopyridin-1-yl)phenyl]pyridin-2-one SMILES: CC1=CN(C(=O)C=C1)c2ccc(cc2)N3C=C(C)C=CC3=O