accessibility menu, dialog, popup

UserName

CAS RN 22515-16-8

IUPAC Name: 1,7-diethyl 4,4-difluoroheptanedioate
Synonyms: 1,7-diethyl 4,4-difluoroheptanedioate
Molecular Weight (g/mol): 252.26
Molecular Formula: C11H18F2O4
InChi Key: XUOBBVMKXUPPEW-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(F)(F)CCC(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)