accessibility menu, dialog, popup

UserName

CAS RN 2252-63-3

IUPAC Name: 1-(4-fluorophenyl)piperazine
Synonyms: para-fluorophenylpiperazine
Molecular Weight (g/mol): 180.23
Molecular Formula: C10H13FN2
InChi Key: AVJKDKWRVSSJPK-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N1CCNCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results