accessibility menu, dialog, popup

UserName

CAS RN 2254-88-8

IUPAC Name: dichloro-1,2,4-thiadiazole
Synonyms: dichloro-1,2,4-thiadiazole
Molecular Weight (g/mol): 155
Molecular Formula: C2Cl2N2S
InChi Key: CEUHPOVLEQUFCC-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=NS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)