CAS RN 226932-59-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl Salicylate Beta-D-O-Glucuronide Methyl Ester, TRC
CAS: 226932-59-8 Molecular Formula: C15H18O9 Molecular Weight (g/mol): 342.3 Synonym: 2-(μethoxycarbonyl)phenyl beta-D-Glucopyranosiduronic alphacid μethyl epsilonster IUPAC Name: methyl (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-methoxycarbonylphenoxy)oxane-2-carboxylate SMILES: COC(=O)[C@H]1O[C@@H](Oc2ccccc2C(=O)OC)[C@H](O)[C@@H](O)[C@@H]1O