accessibility menu, dialog, popup

UserName

CH3(R)O(S)CH3HNH3C

CAS RN 22694-96-8

IUPAC Name: (2R,5S)-2,4,5-trimethyl-2,5-dihydro-1,3-oxazol-3-ium
Synonyms: (2R,5S)-2,4,5-trimethyl-2,5-dihydro-1,3-oxazol-3-ium
Molecular Weight (g/mol): 114.17
Molecular Formula: C6H12NO
InChi Key: YFSGRMONVCFYTC-NTSWFWBYSA-O
SMILES: C[C@H]1O[C@@H](C)C(C)=[NH+]1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results