accessibility menu, dialog, popup

UserName

CAS RN 226942-29-6

HNBr

CAS RN 226942-29-6

IUPAC Name: 6-bromo-1,2,3,4-tetrahydroisoquinoline
Synonyms: 6-bromo-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol): 212.09
Molecular Formula: C9H10BrN
InChi Key: URDGCPQHZSDBRG-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(CNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results