accessibility menu, dialog, popup

UserName

H3COHNOS

CAS RN 22726-30-3

IUPAC Name: 7-methoxy-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 7-methoxy-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: ZITGVVWIRFDIBZ-UHFFFAOYSA-N
SMILES: COC1=CC2=C(NC(=O)CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results