accessibility menu, dialog, popup

UserName

CAS RN 2273-43-0

IUPAC Name: butyl(hydroxy)stannanone
Synonyms: stannane, butylhydroxyoxo-
Molecular Weight (g/mol): 208.83
Molecular Formula: C4H10O2Sn
InChi Key: WIHMDCQAEONXND-UHFFFAOYSA-M
SMILES: CCCC[Sn](O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results